Learn More
MSC GF Gage Thread Plug Gage, 1-3/8-12 Thread, 2B and 3B Class, Hardened Tool Steel, 1 ea
Catalog No. 5000548015
Encompass
This single-end go plug thread gage is designed for the inspection of internal threads, ensuring they meet specified size and class requirements. Constructed from hardened tool steel, this instrument provides reliable thread verification for industrial applications.
  • Thread size is 1-3/8-12
  • Compatible with 2B and 3B classification
  • Hardened tool steel construction
  • Single-end configuration
  • Go pitch diameter is 1.3209 in.
  • Requires size 5 handle (handle not included)
Catalog No. 50-005-48015 Supplier MSC Supplier No. 55307193
This single-end go plug thread gage is designed for the inspection of internal threads, ensuring they meet specified size and class requirements. Constructed from hardened tool steel, this instrument provides reliable thread verification for industrial applications.
  • Thread size is 1-3/8-12
  • Compatible with 2B and 3B classification
  • Hardened tool steel construction
  • Single-end configuration
  • Go pitch diameter is 1.3209 in.
  • Requires size 5 handle (handle not included)