Learn More
MSC GF Gage Plug Thread Gage, #2-64, 3B, No Go, 1 ea
Catalog No. 5000547930
Encompass
This single end thread plug gage is designed for precise inspection of #2-64 threads with a 3B class classification. Engineered for industrial metrology applications, this tool provides a reliable No Go verification for thread pitch diameter.
  • Thread size: #2-64
  • Classification: 3B
  • Type: No Go
  • No go pitch diameter: 0.0779 in.
  • Design: Single end
  • Material: Hardened tool steel
  • Country of origin: USA
Catalog No. 50-005-47930 Supplier MSC Supplier No. 55308498
This single end thread plug gage is designed for precise inspection of #2-64 threads with a 3B class classification. Engineered for industrial metrology applications, this tool provides a reliable No Go verification for thread pitch diameter.
  • Thread size: #2-64
  • Classification: 3B
  • Type: No Go
  • No go pitch diameter: 0.0779 in.
  • Design: Single end
  • Material: Hardened tool steel
  • Country of origin: USA