Learn More
MSC GF GAGE Plug Thread Gage, 3/8-24, 3B, Double End, 1 ea.
Catalog No. 5000511972
Encompass
This double-end plug thread gage is designed for precision inspection of 3/8-24 threads, meeting 3B class requirements. It features a Go and No Go configuration to verify thread accuracy and ensures reliable quality control in machining and manufacturing applications.
  • High speed steel construction for durability
  • TiN coating provides increased wear resistance
  • Double-end design with included handle
  • Includes go and no go gauging members
  • Size: 3/8-24
  • Go pitch diameter: 0.3479 in.
  • No go pitch diameter: 0.3516 in.
  • Classification: 3B
Catalog No. 50-005-11972 Supplier MSC Supplier No. 55314710
This double-end plug thread gage is designed for precision inspection of 3/8-24 threads, meeting 3B class requirements. It features a Go and No Go configuration to verify thread accuracy and ensures reliable quality control in machining and manufacturing applications.
  • High speed steel construction for durability
  • TiN coating provides increased wear resistance
  • Double-end design with included handle
  • Includes go and no go gauging members
  • Size: 3/8-24
  • Go pitch diameter: 0.3479 in.
  • No go pitch diameter: 0.3516 in.
  • Classification: 3B