Learn More
MSC GF Gage Plug Thread Gage, 1/2-20 Thread, 3B Class, No Go, 0.4717 in. Pitch Diameter, 1 ea.
Catalog No. 5000511996
Encompass
This single-end No Go plug thread gage is designed for precision inspection of internal threads. Constructed from high-speed steel, it ensures durability and consistent performance in demanding measurement applications.
  • Thread size of 1/2-20
  • Class 3B classification
  • No Go pitch diameter of 0.4717 in.
  • Single end configuration
  • Constructed from high-speed steel
  • Manufactured in the USA
Catalog No. 50-005-11996 Supplier MSC Supplier No. 55313910
This single-end No Go plug thread gage is designed for precision inspection of internal threads. Constructed from high-speed steel, it ensures durability and consistent performance in demanding measurement applications.
  • Thread size of 1/2-20
  • Class 3B classification
  • No Go pitch diameter of 0.4717 in.
  • Single end configuration
  • Constructed from high-speed steel
  • Manufactured in the USA