Learn More
MSC GF Gage Thread Plug Gage, #8-32, 2B, High-Speed Steel, No Go, 1 ea
Catalog No. 5000511453
Encompass
GF Gage Thread Plug Gage, #8-32, 2B, High-Speed Steel, No Go, 1 ea
Catalog No. 50-005-11453 Supplier MSC Supplier No. 55313175
GF Gage Thread Plug Gage, #8-32, 2B, High-Speed Steel, No Go, 1 ea