Learn More
MSC GF Gage Thread Plug Gage, 1/2-20, 2B, Titanium Nitride, 1 ea
Catalog No. 5000511455
Encompass
This single-end no go thread plug gage is designed for precise inspection of internal threads. Constructed from high-speed steel with a titanium nitride coating, this tool provides durability and wear resistance in demanding industrial environments.
  • Thread size: 1/2-20
  • Class: 2B
  • No go pitch diameter: 0.4731 in.
  • Single end design
  • Titanium nitride finish for enhanced surface hardness
  • High-speed steel construction
Catalog No. 50-005-11455 Supplier MSC Supplier No. 55313472
This single-end no go thread plug gage is designed for precise inspection of internal threads. Constructed from high-speed steel with a titanium nitride coating, this tool provides durability and wear resistance in demanding industrial environments.
  • Thread size: 1/2-20
  • Class: 2B
  • No go pitch diameter: 0.4731 in.
  • Single end design
  • Titanium nitride finish for enhanced surface hardness
  • High-speed steel construction