Learn More
MSC GF Gage Thread Plug Gage, #2-64, 2B and 3B Class, Single End, Go, 1 ea
Catalog No. 5000515011
Encompass
This single-end thread plug gage is designed for precision inspection of #2-64 internal threads. Constructed from hardened tool steel, this go-type gage provides reliable verification for 2B and 3B thread classifications.
  • Hardened tool steel construction for durability
  • Designed for #2-64 thread size
  • Compatible with 2B and 3B class threads
  • Single-end configuration for go inspection
  • Go pitch diameter of 0.0759 in.
Catalog No. 50-005-15011 Supplier MSC Supplier No. 55306179
This single-end thread plug gage is designed for precision inspection of #2-64 internal threads. Constructed from hardened tool steel, this go-type gage provides reliable verification for 2B and 3B thread classifications.
  • Hardened tool steel construction for durability
  • Designed for #2-64 thread size
  • Compatible with 2B and 3B class threads
  • Single-end configuration for go inspection
  • Go pitch diameter of 0.0759 in.