Learn More
MSC GF Gage Thread Plug Gage, 1/2-20, High-Speed Steel, 2B and 3B Class, Go, 1 ea
Catalog No. 5000514577
Encompass
This single-end thread plug gage is designed for precision inspection of 1/2-20 threads. Constructed from high-speed steel with a titanium nitride coating, it offers enhanced durability for rigorous measuring and inspecting applications. This go gage supports both 2B and 3B classes for versatile thread verification.
  • Thread size: 1/2-20
  • Classification: 2B and 3B
  • Go pitch diameter: 0.4675 in.
  • Material: High-speed steel
  • Finish: Titanium nitride
  • Configuration: Single end
  • Handle size: 2
  • Country of origin: United States
Catalog No. 50-005-14577 Supplier MSC Supplier No. 55313035
This single-end thread plug gage is designed for precision inspection of 1/2-20 threads. Constructed from high-speed steel with a titanium nitride coating, it offers enhanced durability for rigorous measuring and inspecting applications. This go gage supports both 2B and 3B classes for versatile thread verification.
  • Thread size: 1/2-20
  • Classification: 2B and 3B
  • Go pitch diameter: 0.4675 in.
  • Material: High-speed steel
  • Finish: Titanium nitride
  • Configuration: Single end
  • Handle size: 2
  • Country of origin: United States