Learn More
MSC GF Gage Thread Plug Gage, 1/2-13, 3B, No Go, 1 ea.
Catalog No. 5000513897
Encompass
This thread plug gage is designed for inspecting internal threads, specifically for a 1/2-13 thread size with a 3B class classification. It is a single-end, no-go gage constructed from hardened tool steel to ensure durability and measurement precision.
  • Thread size: 1/2-13
  • Classification: 3B
  • Type: No-go
  • Design: Single end
  • Material: Hardened tool steel
  • No-go pitch diameter: 0.45480 in.
  • Handle size: 2
Catalog No. 50-005-13897 Supplier MSC Supplier No. 55309033
This thread plug gage is designed for inspecting internal threads, specifically for a 1/2-13 thread size with a 3B class classification. It is a single-end, no-go gage constructed from hardened tool steel to ensure durability and measurement precision.
  • Thread size: 1/2-13
  • Classification: 3B
  • Type: No-go
  • Design: Single end
  • Material: Hardened tool steel
  • No-go pitch diameter: 0.45480 in.
  • Handle size: 2