Learn More
MSC GF Gage Truncated Taperlock Thread Gage, M14 x 2.00, Go, 6G, 3 in. Handle, 1 ea.
Catalog No. 5000511552
Encompass
This truncated taperlock thread gage is designed for precise measurement of M14 x 2.00 metric threads. It is configured as a Go gage for verifying thread accuracy and fits a 3 in. handle for reliable manual operation in inspection environments.
  • Designed for M14 x 2.00 metric threads
  • Configured as a Go gage
  • Features 6G classification
  • Includes a 3 in. handle
  • Manufactured in the USA
Catalog No. 50-005-11552 Supplier MSC Supplier No. 55335491
This truncated taperlock thread gage is designed for precise measurement of M14 x 2.00 metric threads. It is configured as a Go gage for verifying thread accuracy and fits a 3 in. handle for reliable manual operation in inspection environments.
  • Designed for M14 x 2.00 metric threads
  • Configured as a Go gage
  • Features 6G classification
  • Includes a 3 in. handle
  • Manufactured in the USA