Learn More
Essendant TAPETRANSADHES1/2X36YD
Supplier:  Essendant MMM92412
Encompass_Preferred
A great go-to general purpose adhesive transfer film tape with an acrylic reverse wound pressure sensitive adhesive on a densified kraft paper liner Holds- Up to 0 5 lb Permanent/Removable/Repositionable- Permanent Mounting Application- Double-Sided Adhesive Width- 0 5
Catalog No. 50-000-31105