Learn More
Carbosynth LLC. TAPI-0, 5mg, MW: 456.55, Molecular formula: C24H32N4O5, CAS# 143457-40-3
SDP
Supplier:  Carbosynth LLC. INH3850PI5MG
Encompass_Preferred
Synonyms: N-(R)-(2-(Hydroxyaminocarbonyl)Methyl)-4-Methylpentanoyl-L-3-(2'-naphthyl)alanyl-l-alanine amide, HONHCOCH2CH(CH2CH(CH3)2)CO-Nal-Ala-NH2, HONHCOCH2CH(CH2CH(CH3)2)CO-Nal-Ala-NH2, Application: Inhibitor for Collagenase, MMP, and α-Tumor Necrosis Factor, Storage Temperature: -20°C, References: K. Darlak, R.B. Miller, M.S. Stack, A.F. Spatola, and R.D. Grey, J. Biol. Chem., 265 (9), 5199 (1990)., A.F. Spatola, K. Darlak, S. Pegoraro, K. Nijhawan, M. Anzolin, L. Lankiewicz, and R.D. Gray, Peptides: Chemistry & Biology, J.A. Smith & J.E. Rivier (Eds.), ESCOM, Leiden, 1992, p. 820., K.M. Mohler, P.R. Sleath, J.N. Fitzner, D.P. Cerretti, M.Alderson, S.S Kerwar, D.S. Torrance, C. Otten-Evans, T. Greenstreet, K. Weerawarna, S.R. Kronheim, M. Petersen, M. Gerhart, C.J. Kozlosky, C.J. March, and R.A. Black, Nature, 370, 218 (1984).
Catalog No. 50-168-5880