Learn More
Apexbio Technology LLC TAPI-1, 171235-71-5, MFCD02684280, 1 mg
SDP
Supplier:  Apexbio Technology LLC B46861
Encompass_Preferred
MF: C26H37N5O5, MW: 499.6, Solubility: 24.98mg/mL in DMSO. TAPI-1 is an inhibitor of tumour necrosis factor with IC50 value of 8.09 uM.
Catalog No. 50-101-2261