Learn More
Enzo Life Sciences TAPI-2 (1 mg)
SDP
Catalog No. NC0245183
Encompass_Preferred
Inhibitor of TNF-α processing. Inhibitor of TACE, other ADAMs, ACE secretase and other metalloproteinases. Blocks release of TNFα, TGF-α, DCC, APP, L-selectin, ACE, IL6R, NOTCH, and erbB2/HER2. Has been used in culture (typical concentration 5-100µM). Alternative Name: IC3. N-(R)-(2-(Hydroxyaminocarbonyl)methyl)-4-methylpentanoyl-L-t-butyl-glycine-L-alanine 2-aminoethyl amide. Formula: C19H37N5O5. MW: 415.28. Purity: ≥99% (HPLC). Sequence (IC-3). N-(R)-(2-(Hydroxyaminocarbonyl)methyl)-4-methylpentanoyl-L-t-butyl-glycine-L-alanine 2-aminoethyl amide. Appearance: Lyophilized solid. Solubility: Soluble in water (at least 20mM) or 100% ethanol (at least 5mg/ml). Long Term Storage: -20°C
Catalog No. NC0245183 Supplier Enzo Life Sciences Supplier No. BMLPI1350001
Inhibitor of TNF-α processing. Inhibitor of TACE, other ADAMs, ACE secretase and other metalloproteinases. Blocks release of TNFα, TGF-α, DCC, APP, L-selectin, ACE, IL6R, NOTCH, and erbB2/HER2. Has been used in culture (typical concentration 5-100µM). Alternative Name: IC3. N-(R)-(2-(Hydroxyaminocarbonyl)methyl)-4-methylpentanoyl-L-t-butyl-glycine-L-alanine 2-aminoethyl amide. Formula: C19H37N5O5. MW: 415.28. Purity: ≥99% (HPLC). Sequence (IC-3). N-(R)-(2-(Hydroxyaminocarbonyl)methyl)-4-methylpentanoyl-L-t-butyl-glycine-L-alanine 2-aminoethyl amide. Appearance: Lyophilized solid. Solubility: Soluble in water (at least 20mM) or 100% ethanol (at least 5mg/ml). Long Term Storage: -20°C