Learn More
Medchemexpress LLC HY-100211 5mg Medchemexpress, TAPI-2 CAS:187034-31-7 Purity:>98%
SDP
Catalog No. 501931754
Encompass_Preferred
Medchemexpress, HY-100211 5mg TAPI-2 CAS:187034-31-7 TAPI-2 (TNF Protease Inhibitor 2) is a broad-spectrum inhibitor of matrix metalloprotease ( MMP ), tumour necrosis factorα-converting enzyme ( TACE ) and a disintegrin and metalloproteinase ( ADAM ), with an IC 50 of 20±10 μM for MMP. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1754 Supplier Medchemexpress LLC Supplier No. HY1002115MG
Medchemexpress, HY-100211 5mg TAPI-2 CAS:187034-31-7 TAPI-2 (TNF Protease Inhibitor 2) is a broad-spectrum inhibitor of matrix metalloprotease ( MMP ), tumour necrosis factorα-converting enzyme ( TACE ) and a disintegrin and metalloproteinase ( ADAM ), with an IC 50 of 20±10 μM for MMP. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.