Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbosynth LLC. TAPI-2, 1mg, MW: 415.54, Molecular formula: C19H37N5O5, CAS# 689284-12-6
SDP

Catalog No. 501685881
Encompass_Preferred

Synonyms: N-(R)-(2-(Hydroxyaminocarbonyl) Methyl)-4-Methylpentanoyl-L-t-Butyl-Gly-L-Alanine 2-Aminoethyl AmideHONHCOCH2CH(CH2CH(CH3)2)CO-t-Butyl-Gly-Ala-NHCH2CH2NH2, HONHCOCH2CH(CH2CH(CH3)2)CO-t-Butyl-Gly-Ala-NHCH2CH2NH2, Application: Inhibitor for Metalloproteases, Storage Temperature: -20°C, References: N.M. Hooper, E.H. Karran, and A.J. Turner, Biochem. J., 321, 265 (1997).

Catalog No. 50-168-5881 Supplier Carbosynth LLC. Supplier No. INH3852PI1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Synonyms: N-(R)-(2-(Hydroxyaminocarbonyl) Methyl)-4-Methylpentanoyl-L-t-Butyl-Gly-L-Alanine 2-Aminoethyl AmideHONHCOCH2CH(CH2CH(CH3)2)CO-t-Butyl-Gly-Ala-NHCH2CH2NH2, HONHCOCH2CH(CH2CH(CH3)2)CO-t-Butyl-Gly-Ala-NHCH2CH2NH2, Application: Inhibitor for Metalloproteases, Storage Temperature: -20°C, References: N.M. Hooper, E.H. Karran, and A.J. Turner, Biochem. J., 321, 265 (1997).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.