Learn More
Medchemexpress LLC HY-109044 10mg Medchemexpress, Tapinarof CAS:79338-84-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10904410MG
Encompass_Preferred
Medchemexpress, HY-109044 10mg Tapinarof CAS:79338-84-4 Tapinarof (WBI-1001) is a natural aryl hydrocarbon receptor (AhR) agonist with an EC50 of 13 nM. Tapinarof resolves skin inflammation in mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8790