Learn More
Medchemexpress LLC HY-109044 5mg Medchemexpress, Tapinarof CAS:79338-84-4 Purity:>98%
SDP
Catalog No. 501968658
Encompass_Preferred
Medchemexpress, HY-109044 5mg Tapinarof CAS:79338-84-4 Tapinarof (Benvitimod; WBI 1001; GSK2894512) is a natural aryl hydrocarbon receptor (AhR) agonist with an EC50 of 13 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8658 Supplier Medchemexpress LLC Supplier No. HY1090445MG
Medchemexpress, HY-109044 5mg Tapinarof CAS:79338-84-4 Tapinarof (Benvitimod; WBI 1001; GSK2894512) is a natural aryl hydrocarbon receptor (AhR) agonist with an EC50 of 13 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.