Learn More
Medchemexpress LLC Taplucainium chloride | 2489565-37-7 | 98.0% | 386.96 | 100 MG
SDP
Catalog No. 5000361736
Encompass_Preferred
Taplucainium chloride is a sodium channel blocker. It exhibits 70-95% inhibition at 10μM and can be used as an analgesic agent.
  • Sodium channel blocker
  • 70-95% inhibition at 10μM
  • Potential analgesic agent
Catalog No. 50-003-61736 Supplier Medchemexpress LLC Supplier No. HY148802100MG
Taplucainium chloride is a sodium channel blocker. It exhibits 70-95% inhibition at 10μM and can be used as an analgesic agent.
  • Sodium channel blocker
  • 70-95% inhibition at 10μM
  • Potential analgesic agent