Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Taplucainium chloride | 2489565-37-7 | 98.0% | 386.96 | 50 MG
SDP

Catalog No. 5000361526
Encompass_Preferred

Taplucainium chloride is a Sodium Channel blocker that can be used as an analgesic agent. It exhibits 70-95% inhibition at 10μM.

  • Sodium channel blocker
  • Exhibits 70-95% inhibition at 10μM
  • Can be used as an analgesic agent
  • For research use only
  • Available in various sizes
  • Purity of 98.0%
  • Storage: Powder at -20°C for 3 years; in solvent at -80°C for 6 months or -20°C for 1 month
  • Appearance: Solid, off-white to light yellow

Catalog No. 50-003-61526 Supplier Medchemexpress LLC Supplier No. HY14880250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Taplucainium chloride is a Sodium Channel blocker that can be used as an analgesic agent. It exhibits 70-95% inhibition at 10μM.

  • Sodium channel blocker
  • Exhibits 70-95% inhibition at 10μM
  • Can be used as an analgesic agent
  • For research use only
  • Available in various sizes
  • Purity of 98.0%
  • Storage: Powder at -20°C for 3 years; in solvent at -80°C for 6 months or -20°C for 1 month
  • Appearance: Solid, off-white to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.