Learn More
Medchemexpress LLC Taplucainium chloride | 2489565-37-7 | 98.0% | 386.96 | C23H31ClN2O | 10 MG
SDP
Catalog No. 5000285711
Encompass_Preferred
Taplucainium chloride is a small-molecule sodium channel blocker intended for laboratory research. It exhibits 70-95% inhibition of sodium channel activity at 10 μM and is reported to have analgesic potential. The compound is supplied as an off-white to light-yellow solid with a reported purity of 98.0% and is available as a solid or solution for experimental use.
  • Small-molecule sodium channel blocker with demonstrated activity (70-95% inhibition at 10 μM).
  • Reported purity of 98.0%.
  • Available as a solid and as a 10 mM solution in DMSO for convenience.
  • Off-white to light yellow solid appearance suitable for research applications.
  • Recommended storage: powder at -20 °C; solutions at -80 °C (6 months) or -20 °C (1 month).
  • For research use only; not for clinical or human use.
Catalog No. 50-002-85711 Supplier Medchemexpress LLC Supplier No. HY14880210MG
Taplucainium chloride is a small-molecule sodium channel blocker intended for laboratory research. It exhibits 70-95% inhibition of sodium channel activity at 10 μM and is reported to have analgesic potential. The compound is supplied as an off-white to light-yellow solid with a reported purity of 98.0% and is available as a solid or solution for experimental use.
  • Small-molecule sodium channel blocker with demonstrated activity (70-95% inhibition at 10 μM).
  • Reported purity of 98.0%.
  • Available as a solid and as a 10 mM solution in DMSO for convenience.
  • Off-white to light yellow solid appearance suitable for research applications.
  • Recommended storage: powder at -20 °C; solutions at -80 °C (6 months) or -20 °C (1 month).
  • For research use only; not for clinical or human use.