Learn More
Medchemexpress LLC Taprenepag isopropyl | 1005549-94-9 | 99.0% | 520.60 | 1 ML
SDP
Catalog No. 5000405069
Encompass_Preferred
Taprenepag isopropyl is a highly selective EP2 receptor agonist. It reduces intraocular pressure (IOP) in the right (dosed) eye in a dose-dependent manner in cynomolgus monkeys. In a high-dose group (0.75 mg/day), IOP was reduced to less than 4 mm Hg on Days 22 and 29. No clinical signs or changes in body weight were observed with any dose, and no ocular findings occurred in the low-dose group (0.75 mg/day).
  • Highly selective EP2 receptor agonist.
  • Reduces intraocular pressure (IOP) in a dose-dependent manner.
  • Achieved IOP reduction to less than 4 mm Hg in high-dose group.
  • No clinical signs or changes in body weight observed.
  • No ocular findings occurred in the low-dose group.
  • Target: EP2.
Catalog No. 50-004-05069 Supplier Medchemexpress LLC Supplier No. HY1999810MM/1ML
Taprenepag isopropyl is a highly selective EP2 receptor agonist. It reduces intraocular pressure (IOP) in the right (dosed) eye in a dose-dependent manner in cynomolgus monkeys. In a high-dose group (0.75 mg/day), IOP was reduced to less than 4 mm Hg on Days 22 and 29. No clinical signs or changes in body weight were observed with any dose, and no ocular findings occurred in the low-dose group (0.75 mg/day).
  • Highly selective EP2 receptor agonist.
  • Reduces intraocular pressure (IOP) in a dose-dependent manner.
  • Achieved IOP reduction to less than 4 mm Hg in high-dose group.
  • No clinical signs or changes in body weight observed.
  • No ocular findings occurred in the low-dose group.
  • Target: EP2.