Learn More
Medchemexpress LLC Taprenepag isopropyl | 1005549-94-9 | 99.0% | 520.60 | 50 MG
SDP
Catalog No. 5000404402
Encompass_Preferred
Taprenepag isopropyl is a highly selective EP2 receptor agonist. In preclinical studies, administration of Taprenepag isopropyl led to a reduction in intraocular pressure (IOP) in dose-dependent manner. Clinical trials have investigated its potential in conditions such as Primary Open-Angle Glaucoma and Ocular Hypertension.
  • Highly selective receptor interaction
  • Demonstrates significant reduction in intraocular pressure in animal models
  • Evaluated in clinical trials for specific ocular conditions
  • Solid, white to off-white appearance
Catalog No. 50-004-04402 Supplier Medchemexpress LLC Supplier No. HY1999850MG
Taprenepag isopropyl is a highly selective EP2 receptor agonist. In preclinical studies, administration of Taprenepag isopropyl led to a reduction in intraocular pressure (IOP) in dose-dependent manner. Clinical trials have investigated its potential in conditions such as Primary Open-Angle Glaucoma and Ocular Hypertension.
  • Highly selective receptor interaction
  • Demonstrates significant reduction in intraocular pressure in animal models
  • Evaluated in clinical trials for specific ocular conditions
  • Solid, white to off-white appearance