Learn More
TARGETMOL CHEMICALS INC TAPRIZOSIN 5MG
SDP
Catalog No. 502580477
Encompass_Preferred
Also available in 50 mg 100 mg and bulk. Please contact Fisher for quotes. Taprizosin(UK-338003) is a selective and orally active (alpha)1-adrenoceptor antagonist for vasodilatation in the treatment of benign prostatic hyperplasia. purity: 99%
Catalog No. 50-258-0477 Supplier TARGETMOL CHEMICALS INC Supplier No. T289225MG
Also available in 50 mg 100 mg and bulk. Please contact Fisher for quotes. Taprizosin(UK-338003) is a selective and orally active (alpha)1-adrenoceptor antagonist for vasodilatation in the treatment of benign prostatic hyperplasia. purity: 99%