Learn More
Research Products International Corp TAPS 1 KG
SDP
Catalog No. 501977912
Encompass_Preferred
pKa(25°:C): 8.4. The working pH range in aqueous solution is 7.7-9.1.
Catalog No. 50-197-7912 Supplier Research Products International Corp Supplier No. T120001000.0
pKa(25°:C): 8.4. The working pH range in aqueous solution is 7.7-9.1.