Learn More
Research Products International Corp TAPS, 100 G
SDP
Catalog No. 501978436
Encompass_Preferred
pKa(25°:C): 8.4. The working pH range in aqueous solution is 7.7-9.1.
Catalog No. 50-197-8436 Supplier Research Products International Corp Supplier No. T12000100.0
pKa(25°:C): 8.4. The working pH range in aqueous solution is 7.7-9.1.