Learn More
Research Products International Corp TAPS, 5 KG
SDP
Catalog No. 501977186
Encompass_Preferred
pKa(25°:C): 8.4. The working pH range in aqueous solution is 7.7-9.1.
Catalog No. 50-197-7186 Supplier Research Products International Corp Supplier No. T120005000.0
pKa(25°:C): 8.4. The working pH range in aqueous solution is 7.7-9.1.