Learn More
Research Products International Corp TAPS, [Tris(hydroxymethyl)-3-aminopropanesulfonic acid], Free Acid, 500 Grams
SDP
Catalog No. 501368615
Encompass_Preferred
pKa(25°:C): 8.4. The working pH range in aqueous solution is 7.7-9.1.
Catalog No. 50-136-8615 Supplier Research Products International Corp Supplier No. T12000500.0
pKa(25°:C): 8.4. The working pH range in aqueous solution is 7.7-9.1.