Learn More
Medchemexpress LLC Tapso | 68399-81-5 | 98.0% | 100 G
SDP
Catalog No. 5000410456
Encompass_Preferred
TAPSO is a widely used biological buffer and pH stabilization reagent. It offers an effective pH range between 7 and 8, with pKa values typically falling between 7.5 and 9.0. This compound contains Tris groups and has been observed to exhibit reactive activity with zwitterionic glycine peptides. It is primarily intended for research applications.
  • Effective pH range of 7-8.
  • pKa values ranging from 7.5-9.0.
  • Contains Tris groups.
  • Shows reactive activity with zwitterionic glycine peptides.
  • Suitable for various research applications.
Catalog No. 50-004-10456 Supplier Medchemexpress LLC Supplier No. HYD0878100G
TAPSO is a widely used biological buffer and pH stabilization reagent. It offers an effective pH range between 7 and 8, with pKa values typically falling between 7.5 and 9.0. This compound contains Tris groups and has been observed to exhibit reactive activity with zwitterionic glycine peptides. It is primarily intended for research applications.
  • Effective pH range of 7-8.
  • pKa values ranging from 7.5-9.0.
  • Contains Tris groups.
  • Shows reactive activity with zwitterionic glycine peptides.
  • Suitable for various research applications.