Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Tapwater Tour
SDPSCI_ED

Catalog No. S19119R Shop All LaMotte Company Products
Change view
Click to view available options
Refill Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S19119R Refill Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S19119R Supplier LaMotte™ Supplier No. R3608

missing translation for 'validMainframeMsg'

Includes

Lecture materials, illustrated handouts, teacher tips, test procedures and wrap-up games

Reagent refill package consisting of a set of consumable items as originally supplied in the kit.

  • Uses simple and safe reagent tablets in unit-dose form
  • Includes five units: pH, chlorine, hardness, iron and copper

Order Info

Grades 4 to 6. Class size: 50 students.

Specifications

For Use With (Application) Analyze the quality of tap water at home or in the classroom
Grade 4 to 6
Includes Lecture materials, illustrated handouts, teacher tips, test procedures and wrap up games
Sufficient For 50 Students
Type Refill Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.