Learn More
New England Biolabs, Inc. Taq 5X Master Mix – 500 reactions (50 µl vol)
SDP
Catalog No. 50995179
Encompass
Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5' to 3' polymerase activity and a 5' flap endonuclease activity. Taq 5X Master Mix is an optimized ready-to-use solution containing Taq DNA Polymerase, dNTPs, MgCl2, KCl and stabilizers. It is best for routine PCR applications from templates including pure DNA solutions, bacterial colonies, and cDNA. It can amplify up to 4 kb from complex genomic DNA or up to 5 kb from lambda DNA.
Catalog No. 50-995-179 Supplier New England Biolabs, Inc. Supplier No. M0285L
Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5' to 3' polymerase activity and a 5' flap endonuclease activity. Taq 5X Master Mix is an optimized ready-to-use solution containing Taq DNA Polymerase, dNTPs, MgCl2, KCl and stabilizers. It is best for routine PCR applications from templates including pure DNA solutions, bacterial colonies, and cDNA. It can amplify up to 4 kb from complex genomic DNA or up to 5 kb from lambda DNA.