Learn More
Qiagen Beverly Inc TAQ-B DNA POLYMERASE
Catalog No. 50305926
Encompass
DNA Polymerase; Taq-B DNA Polymerase; Conc. 5,000 U/ml; 10,000 U; incl. 10X buffer
Catalog No. 50-305-926 Supplier Qiagen Beverly Inc Supplier No. P7250L
DNA Polymerase; Taq-B DNA Polymerase; Conc. 5,000 U/ml; 10,000 U; incl. 10X buffer