Learn More
Meridian Life Science Taq Dilution Buffer, 5ML
SDP
Catalog No. 5000856388
Encompass_Preferred
Taq Dilution Buffer is an optimized buffer system that has been designed to conferring long-term stability at -20 C to high-quality Taq DNA polymerase such as MDX001 or MDX008. The reaction mix requires addition of Reaction Buffer, dNTPs and MgCl2, together with primers and template.
Catalog No. 50-008-56388 Supplier Meridian Life Science Supplier No. MDX0075
Taq Dilution Buffer is an optimized buffer system that has been designed to conferring long-term stability at -20 C to high-quality Taq DNA polymerase such as MDX001 or MDX008. The reaction mix requires addition of Reaction Buffer, dNTPs and MgCl2, together with primers and template.