Learn More
New England Biolabs, Inc. Taq DNA Ligase – 10000 units
SDP
Catalog No. 50811617
Encompass
Taq DNA Ligase is a thermostable ligase that catalyzes the formation of a phosphodiester bond between the 5'-phosphate and the 3'-hydroxyl of two adjacent DNA strands. The strands to be ligated need to be hybridized and accurately paired, with no gap, to a complementary DNA strand; allowing resolution of single nucleotide variants. Taq DNA Ligase uses NAD as a cofactor and it is active at elevated temperatures (37 C - 75 C).
Catalog No. 50-811-617 Supplier New England Biolabs, Inc. Supplier No. M0208L
Taq DNA Ligase is a thermostable ligase that catalyzes the formation of a phosphodiester bond between the 5'-phosphate and the 3'-hydroxyl of two adjacent DNA strands. The strands to be ligated need to be hybridized and accurately paired, with no gap, to a complementary DNA strand; allowing resolution of single nucleotide variants. Taq DNA Ligase uses NAD as a cofactor and it is active at elevated temperatures (37 C - 75 C).