Learn More
New England Biolabs, Inc. Taq DNA Ligase Reaction Buffer – 6 ml
SDP
Catalog No. 50811512
Encompass
This is the NEB reaction buffer supplied with Taq DNA Ligase. Taq DNA Ligase requires NAD+ as a cofactor which is incorporated into the buffer. New England Biolabs supplies a 10X reaction buffer with all of its enzymes. At a 1X concentration this reaction buffer assures optimal activity of the enzyme.
Catalog No. 50-811-512 Supplier New England Biolabs, Inc. Supplier No. B0208S
This is the NEB reaction buffer supplied with Taq DNA Ligase. Taq DNA Ligase requires NAD+ as a cofactor which is incorporated into the buffer. New England Biolabs supplies a 10X reaction buffer with all of its enzymes. At a 1X concentration this reaction buffer assures optimal activity of the enzyme.