Learn More
Qiagen Beverly Inc TAQ DNA LIGASE
Catalog No. 50305910
Encompass
DNA Ligase, Taq DNA ligase; Conc. 40,000 U/ul; 20,000U; incl. 10X buffer
Catalog No. 50-305-910 Supplier Qiagen Beverly Inc Supplier No. L6060L
DNA Ligase, Taq DNA ligase; Conc. 40,000 U/ul; 20,000U; incl. 10X buffer