Learn More
INTACT GENOMICS INC Taq DNA Polymerase 2x Premix 500 Reactions
Catalog No. 5000082905
Encompass
IG™ Taq DNA Polymerase 2x Premix with Dye is a thermostable DNA polymerase that possesses a 5´→3´ polymerase activity (1, 2) and a 5´ flap endonuclease activity (3, 4). Ideal for primary extension reaction with DNA fragments having dA overhang on 3’ ends.IG™ Taq DNA Polymerase 2x Premix with Dye is ready to use, containing Taq DNA Polymerase, dNTPs, MgCl2, and stabilizers. It has been optimized for routine PCR applications.Applications PCR Primer extension Colony PCR
Catalog No. 50-000-82905 Supplier INTACT GENOMICS INC Supplier No. 3249
IG™ Taq DNA Polymerase 2x Premix with Dye is a thermostable DNA polymerase that possesses a 5´→3´ polymerase activity (1, 2) and a 5´ flap endonuclease activity (3, 4). Ideal for primary extension reaction with DNA fragments having dA overhang on 3’ ends.IG™ Taq DNA Polymerase 2x Premix with Dye is ready to use, containing Taq DNA Polymerase, dNTPs, MgCl2, and stabilizers. It has been optimized for routine PCR applications.Applications PCR Primer extension Colony PCR