Learn More
New England Biolabs, Inc. Taq DNA Polymerase with Standard Taq Buffer – 2000 units
SDP
Catalog No. 50994876
Encompass
Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5' to 3' polymerase activity and a 5' flap endonuclease activity. It is supplied with 10X Standard Taq Reaction Buffer, which is detergent-free and designed to be compatible with existing assay systems.
Catalog No. 50-994-876 Supplier New England Biolabs, Inc. Supplier No. M0273L
Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5' to 3' polymerase activity and a 5' flap endonuclease activity. It is supplied with 10X Standard Taq Reaction Buffer, which is detergent-free and designed to be compatible with existing assay systems.