Learn More
New England Biolabs, Inc. Taq DNA Polymerase with ThermoPol Buffer – 2000 units
SDP
Catalog No. 50811694
Encompass
Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 53 polymerase activity and a 5 flap endonuclease activity. It is supplied with 10X ThermoPol Reaction Buffer, which contains a nonionic detergent to increase enzyme stability during longer incubations.
Catalog No. 50-811-694 Supplier New England Biolabs, Inc. Supplier No. M0267L
Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 53 polymerase activity and a 5 flap endonuclease activity. It is supplied with 10X ThermoPol Reaction Buffer, which contains a nonionic detergent to increase enzyme stability during longer incubations.