Learn More
Genscript Corporation Taq DNA Polymerase 1000 U wi
Catalog No. 5000058967
Encompass
The Product contain 1000 U Taq DNA Polymerase (Cat No E00007) and 0 5 ml of 10 mM dNTP (Cat No D0056) The Taq DNA Polymerase is supplied with 10X reaction buffer (Cat No B0005) containing 15 mM magnesium chloride
Catalog No. 50-000-58967 Supplier Genscript Corporation Supplier No. E00101
The Product contain 1000 U Taq DNA Polymerase (Cat No E00007) and 0 5 ml of 10 mM dNTP (Cat No D0056) The Taq DNA Polymerase is supplied with 10X reaction buffer (Cat No B0005) containing 15 mM magnesium chloride