Learn More
Takara Bio TaKaRa Taq™ DNA Polymerase, 250 units
SDP
Catalog No. 50443948
Encompass
Takara Taq DNA Polymerase is a recombinant version of full-length Taq polymerase. It has the same characteristics and capabilities as the native Taq polymerase, and is suitable for a variety of standard PCR applications. The polymerase is supplied with separate tubes of buffer (Mg2+ plus) and dNTPs. 250 units
Catalog No. 50-443-948 Supplier Takara Bio Supplier No. R001A
Takara Taq DNA Polymerase is a recombinant version of full-length Taq polymerase. It has the same characteristics and capabilities as the native Taq polymerase, and is suitable for a variety of standard PCR applications. The polymerase is supplied with separate tubes of buffer (Mg2+ plus) and dNTPs. 250 units