Learn More
Sigma Aldrich Fine Chemicals Biosciences Taq DNA Polymerase 5 U/mul
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11596594001
Encompass
Taq DNA Polymerase is a highly processive 5-3 DNA polymerase that lacks 3-5 exonuclease activity. It is a single polypeptide chain with a molecular weight of approximately 95 kDa.Taq DNA Polymerase was originally isolated from the thermophilic eubacterium Thermus aquaticus BM a strain lacking Taq I restriction endonuclease. The enzyme was cloned in E.coli.
Catalog No. 50-291-4712