Learn More
INTACT GENOMICS INC Taq DNA Polymerase 5000 Units
Catalog No. 5000082959
Encompass
Intact Genomics Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5´→3´ polymerase activity (1, 2) and a 5´ flap endonuclease activity (3, 4). This product is supplied with 10x PCR reaction buffer, containing MgCl2, which produces a final Mg2+ concentration of 1.5 mM. Ideal for primary extension reaction with DNA fragments having dA overhang on 3’ ends.Applications Routing PCR cloning Primer extension Colony PCR Elongation efficiency 1.0-1.2 kb/min. Formulated for amplifying long target DNA. Efficient for amplifying high GC content DNA with Intact Genomics magic enhancer
Catalog No. 50-000-82959 Supplier INTACT GENOMICS INC Supplier No. 3245
Intact Genomics Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5´→3´ polymerase activity (1, 2) and a 5´ flap endonuclease activity (3, 4). This product is supplied with 10x PCR reaction buffer, containing MgCl2, which produces a final Mg2+ concentration of 1.5 mM. Ideal for primary extension reaction with DNA fragments having dA overhang on 3’ ends.Applications Routing PCR cloning Primer extension Colony PCR Elongation efficiency 1.0-1.2 kb/min. Formulated for amplifying long target DNA. Efficient for amplifying high GC content DNA with Intact Genomics magic enhancer