Learn More
Genscript Corporation Taq DNA Polymerase
Supplier:  Genscript Corporation E0000750000
Encompass
Taq DNA Polymerase is a thermostable DNA Polymerase isolated from an E. coli strain that carries the Taq DNA polymerase gene. Taq DNA Polymerase is the most common polymerase used for PCR.Note: This product is supplied with 10X reaction buffer containing 15 mM magnesium chloride. dNTP (10 mM) mixture must be ordered separately (see related products).
Catalog No. 50-274-392