Learn More
Sigma Aldrich Fine Chemicals Biosciences Taq DNA Polymerase dNTPack
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 4728904001
Encompass
Taq DNA Polymerase is a highly processive 5-3 DNA polymerase that lacks 3-5 exonuclease activity. It is a single polypeptide chain with a molecular weight of approximately 95 kDa.Taq DNA Polymerase was originally isolated from the thermophilic eubacterium Thermus aquaticus BM a strain lacking Taq I restriction endonuclease. The enzyme preparation obtained from E.coli is free of nonspecific engo- or exonucleases according to the current quality control procedures.
Catalog No. 50-291-4713