Learn More
Meridian Life Science Taq DNA Polymerase Enzyme only, 100ML
SDP
Catalog No. 5000856218
Encompass_Preferred
Taq DNA Polymerase has been developed to give more robust amplification than other polymerases allowing it to perform well with challenging templates in the presence of PCR inhibitors, making it ideal for fast PCR reactions across a range of templates, without compromising specificity or yield.
Catalog No. 50-008-56218 Supplier Meridian Life Science Supplier No. MDX001100A
Taq DNA Polymerase has been developed to give more robust amplification than other polymerases allowing it to perform well with challenging templates in the presence of PCR inhibitors, making it ideal for fast PCR reactions across a range of templates, without compromising specificity or yield.