Learn More
Genscript Corporation Taq DNA Polymerase without Mg
Catalog No. 5000058351
Encompass
DNA Polymerase is a thermostable DNA polymerase isolated from an i E coli /i strain that carries the i Taq /i DNA polymerase gene i Taq /i DNA polymerase is the most common polymerase used for PCR
Catalog No. 50-000-58351 Supplier Genscript Corporation Supplier No. E00008
DNA Polymerase is a thermostable DNA polymerase isolated from an i E coli /i strain that carries the i Taq /i DNA polymerase gene i Taq /i DNA polymerase is the most common polymerase used for PCR