Learn More
Agilent Technologies TAQ EXTENDER PCR ADD.TAQ EXTE
Catalog No. 50125922
Encompass
Taq Extender PCR Additive improves length, yield and reliability of PCR. Enhances efficiency of template extension by Taq DNA polymerase.
Catalog No. 50-125-922 Supplier Agilent Technologies Supplier No. 600148
Taq Extender PCR Additive improves length, yield and reliability of PCR. Enhances efficiency of template extension by Taq DNA polymerase.