Learn More
Takara Bio TAQ HOT START PREMIX
SDP
Catalog No. 50443964
Encompass
PCR Reagents; Takara; Hot-Start Taq DNA Polymerase; 100rxns; reduces background, increased specificity, permits room-temperature reaction assembly
Catalog No. 50-443-964 Supplier Takara Bio Supplier No. R028A
PCR Reagents; Takara; Hot-Start Taq DNA Polymerase; 100rxns; reduces background, increased specificity, permits room-temperature reaction assembly