Learn More
Meridian Life Science Taq HS Antibody, 20UL
SDP
Catalog No. 5000856391
Encompass_Preferred
Taq HS Antibody is a mix of anti-Taq antibodies designed to inhibit Taq DNA polymerase activity at room temperature. It is used in the preparation of hot-start PCR or qPCR master mixes and helps to prevent nonspecific amplification and primer-dimer formation, increasing specificity and sensitivity while allowing assembly of reactions at ambient temperatures. Inhibition is reversed when the temperature is raised during the first denaturing step of thermal cycling.
Catalog No. 50-008-56391 Supplier Meridian Life Science Supplier No. MDX014002
Taq HS Antibody is a mix of anti-Taq antibodies designed to inhibit Taq DNA polymerase activity at room temperature. It is used in the preparation of hot-start PCR or qPCR master mixes and helps to prevent nonspecific amplification and primer-dimer formation, increasing specificity and sensitivity while allowing assembly of reactions at ambient temperatures. Inhibition is reversed when the temperature is raised during the first denaturing step of thermal cycling.